Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2481

Sigma-Aldrich

BI-9321 Hydrochloride

≥98% (HPLC)

Synonym(s):

{4-[5-(7-fluoroquinolin-4-yl)-1-methyl-1H-imidazol-4-yl]-3,5-dimethylphenyl}-methanamine Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21FN4 · xHCl
Molecular Weight:
360.43 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to very dark green-yellow

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

BI-9321 is a highly selective and potent inhibitor of histone lysine methyltransferase NSD3 PWWP-1 domain that binds to methylated Lysine residue.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service