Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01080

Sigma-Aldrich

Semilicoisoflavone B

≥85% (LC/MS-ELSD)

Synonym(s):

5,7,8′-Trihydroxy-2′,2′-dimethyl-2′H-(3,6′)bi(1-benzopyranyl)-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16O6
CAS Number:
Molecular Weight:
352.34
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥85% (LC/MS-ELSD)

form

solid

mol wt

352.34

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[o]1c2c([c](c(c1)c3cc4c(c(c3)O)OC(C=C4)(C)C)=O)c(cc(c2)O)O

InChI

1S/C20H16O6/c1-20(2)4-3-10-5-11(6-15(23)19(10)26-20)13-9-25-16-8-12(21)7-14(22)17(16)18(13)24/h3-9,21-23H,1-2H3

InChI key

LWZACZCRAUQSLH-UHFFFAOYSA-N

General description

Semilicoisoflavone B, is an isoflavonoid belongs to the class of 7-hydroxyisoflavones, is a natural product commonly available from Glycyrrhiza Sp., Mitracarpus scaber, and Lentinus tigrinus. This plant metabolite exhibits various biological activities, including anti-inflammatory, antioxidant, anti-tumor, antiviral, and anti-allergic effects.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Semilicoisoflavone B:
  • Suppresses proinflammatory cytokine production, downregulates TLR4 expression, and diminishes reactive oxygen species generation in vitro.
  • Inhibits the growth and division of cancer cells by triggering apoptosis and arresting the cell cycle, offering potential for effective tumor therapy.
  • Restricts Caco-2 cell proliferation, suggesting potential for metabolic syndrome therapy.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service