Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00108

Sigma-Aldrich

Apigenin 7-glucuronide

≥95% (LC/MS-ELSD)

Synonym(s):

Apigenin 7-O-β-glucuronide, 4′,5,7-Trihydroxyflavone 7-glucuronide, Scutellarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)cc4

InChI

1S/C21H18O11/c22-9-3-1-8(2-4-9)13-7-12(24)15-11(23)5-10(6-14(15)31-13)30-21-18(27)16(25)17(26)19(32-21)20(28)29/h1-7,16-19,21-23,25-27H,(H,28,29)/t16-,17-,18+,19-,21+/m0/s1

InChI key

JBFOLLJCGUCDQP-ZFORQUDYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariarosa Maietta et al.
Food & function, 9(3), 1545-1556 (2018-02-13)
Accumulation of advanced glycation end products (AGEs) in vivo is associated with many chronic disorders such as diabetes, renal failure, aging, and Alzheimer's disease. The aim of this study was to expand the knowledge about the functional properties of Origanum

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service