Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAE0078

Sigma-Aldrich

MMP-9 pre-activated human

recombinant, ≥1,300 pmol/min/μg, expressed in HEK 293 cells

Synonym(s):

GELB, GELBCLG4B, Gelatinase, Gelatinase B, MANDP2, MMP-9, Matrix Metalloproteinase-10, Type IV collagenase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

calculated mol wt 66 kDa
observed mol wt 82 kDa by SDS-PAGE (The protein migrates as a 82 kDa protein on SDS-PAGE due to glycosylation)

concentration

≥50 μg/mL

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MMP(4318)

General description

50 μg protein, determined by Bradford. The protein concentration is ≥50 μg/ml. for the lot-specific concentration, see Certificate of Analysis.

Biochem/physiol Actions

MMP-9 is a member of the matrix metalloproteinase (MMP) family of proteins. Proteins of the MMP family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Studies in rhesus monkeys suggest that MMP9 is involved in IL-8 (interleukin-8)-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling. Thrombospondins, intervertebral disc proteins, regulate the effective levels of MMP-2 and -9, which are key effectors of extracellular matrix (ECM) remodeling. MMP-9 degrades various substrates including gelatin, collagen types IV and V, and elastin. MMP-9 is involved in a variety of autoimmune diseases such as systemic lupus erythematosus, rheumatoid arthritis, and multiple sclerosis, and be regarded as a potential therapeutic target.
As with most MMPs, MMP-9 is secreted as an inactive pro-protein which is activated when cleaved by extracellular proteinases. MMP-9 can be cleaved (and thus activated) in vitro using 4-Aminophenylmercuric acetate (APMA), Cat. No. A9563. MMP-9 is secreted from neutrophils, macrophages, and a number of transformed cells, and is the most complex family member in terms of domain structure and regulation of its activity.
Structurally, MMP9 maybe be divided into five distinct domains: a pro-domain which is cleaved upon activation, a gelatin binding domain consisting of three contiguous fibronectin type II units, a catalytic domain containing the zinc binding site, a proline rich linker region, and a carboxyl terminal hemopexin like domain.
As with most MMPs, MMP-9 is secreted as an inactive pro-protein which is activated when cleaved by extracellular proteinases. This product was cleaved and activated in vitro using 4-Aminophenylmercuric acetate (APMA), Cat. No. A9563.

Other Notes

This product was pre-activated in vitro using 4-Aminophenylmercuric acetate (APMA). Thus, it is active and ready for use. In order to save our customers from handling hazardous materials, and for environmental saving, the highly toxic and fatal APMA was removed from the final preparation.

Physical form

Liquid solution, 0.22 mm filtered, containing 25 mM Tris, 10 mM CaCl2, 0.05% Brij-35

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geraldine M Jowett et al.
Nature materials, 20(2), 250-259 (2020-09-09)
Organoids can shed light on the dynamic interplay between complex tissues and rare cell types within a controlled microenvironment. Here, we develop gut organoid cocultures with type-1 innate lymphoid cells (ILC1) to dissect the impact of their accumulation in inflamed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service