Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5700532

Sigma-Aldrich

Anti-Pan Phospho-Tyrosine antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

concentration

0.86mg/ml

technique(s)

immunofluorescence: 1:50-1:200
western blot: 1:500-1:2000

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pan pTyr)

Immunogen

A synthetic peptide corresponding to a sequence containing phosphorylated Y.

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service