Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB4200781

Sigma-Aldrich

Anti-Haptoglobin antibody, Mouse monoclonal

clone HG-36, purified from hybridoma cell culture

Synonym(s):

Anti-HP, Anti-Zonulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HG-36, monoclonal

form

buffered aqueous solution

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

ELISA: 0.2-0.4 μg/mL using 10 μg/ml human haptoglobin for coating
dot blot: suitable
immunoblotting: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HP(3240)

General description

Haptoglobin, also known as HP, is a serum α2-glycoprotein that exists as a tetramer, composed of two smaller identical alpha (α) chains and two larger identical beta (β) chains. The α-chains are linked to each other by a disulfide bond and each β-chain is similarly linked to an α-chain.
Monoclonal Anti-Haptoglobin specifically recognizes the three types of human haptoglobin (types 1-1, 2-1, and 2-2) The antibody recognizes both purified and haptoglobin in human serum, and shows no crossreactivity with human IgG, transferrin, C-reactive protein, α-1-acid glycoprotein, albumin, hemoglobin, or bovine, goat and sheep serum haptoglobin.

Immunogen

purified haptoglobin from pooled human plasma

Application

The antibody is recommended to use in various immunological techniques, including ELISA, dot blot, immunoprecipitation, and immunoblot . The antibody reacts against native and denatured (non-reduced) human haptoglobin but does not recognize the haptoglobin under reducing conditions.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service