Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2100465

Sigma-Aldrich

Anti-COL6A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Collagen, type VI, α 1, Anti-OPLL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

106 kDa

species reactivity

dog, pig, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL6A1(1291)

Immunogen

Synthetic peptide directed towards the middle region of human COL6A1

Biochem/physiol Actions

COL6A1 is a collagen VI which acts as a cell-binding protein.Defects in COL6A1 are a cause of Bethlem myopathy (BM).The collagens are a superfamily of proteins that play a role in maintaining the integrity of various tissues. Collagens are extracellular matrix proteins and have a triple-helical domain as their common structural element. Collagen VI is a major structural component of microfibrils. The basic structural unit of collagen VI is a heterotrimer of the alpha1(VI), alpha2(VI), and alpha3(VI) chains. The alpha2(VI) and alpha3(VI) chains are encoded by the COL6A2 and COL6A3 genes, respectively. The protein encoded by this gene is the alpha 1 subunit of type VI collagen (alpha1(VI) chain). Mutations in the genes that code for the collagen VI subunits result in the autosomal dominant disorder, Bethlem myopathy. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. This gene, which encodes a neural small non-messenger RNA, is a member of the family of interspersed repetitive DNA, and its product represents an example of a primate tissue-specific RNA polymerase III transcript. The RNA sequence is divided into three domains: a 5′ portion homologous to the Alu Lm, a central adenosine-rich region, and the terminal 43-nt nonrepetitive domain. It is believed that this gene was retropositionally generated and recruited into a function regulating dendritic protein biosynthesis. At least two pseudogenes of this gene have been identified. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-200 AF020057.2 4262-4461

Sequence

Synthetic peptide located within the following region: ADITILLDGSASVGSHNFDTTKRFAKRLAERFLTAGRTDPAHDVRVAVVQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service