Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1405181

Sigma-Aldrich

Monoclonal Anti-NXF2 antibody produced in mouse

clone 4F5-3A2, ascites fluid

Synonym(s):

FLJ20416, TAPL-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4F5-3A2, monoclonal

mol wt

antigen ~94.97 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgMκ

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NXF2(56001)

General description

This gene is one of a family of nuclear RNA export factor genes. It encodes a protein that is involved in mRNA export, is located in the nucleoplasm, and is associated with the nuclear envelope. Alternative splicing seems to be a common mechanism in this gene family. Two variants have been found for this gene. (provided by RefSeq)

Immunogen

NXF2 (AAH15020, 1 a.a. ~ 626 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MCSTLKKCGTYRTEVAECHDHGSTFQGRKKGGSSFRDNFDKRSCHYEHGGYERPPSHCQENDGSVEVRDVHKDQQLRHTPYSIRCERRMKWHSEDEIRITTWRNRKPPERKMGQNTQDGYTRNWFKVTIPYGIKYDKAWLMNSIQSHCSDRFTPVDFHYVRNRACFFVQDASAASALKDVSYKIYDDENQKICIFVNHSTAPYSVKNKLRPGQMEMLKLTMNKRYNVSQQALDLQNLRFDPDLMGRDIDIILNRRNCMAATLKIIERNFPELLSLNLCNNKLYQLDGLSDITEKAPKVKTLNLSKNKLESAWELGKVKGLKLEELWLEGNPLCSTFSDQSAYVSAIRDCFPKLLRLDGRELSAPVIVDIDSSETMKPCKENFTGSETLKHLVLQFLQQYYSIYDSGDRQGLLGAYHDEACFSLAIPFDPKDSAPSSLCKYFEDSRNMKTLKDPYLKGELLRRTKRDIVDSLSALPKTQHDLSSILVDVWCQTERMLCFSVNGVFKEVEGQSQGSVLAFTRTFIATPGSSSSLCIVNDELFVRDASPQETQSAFSIPVSTLSSSSEPSLSQEQQEMVQAFSAQSGMKLEWSQKCLQDNEWNYTRAGQAFTMLQTEGKIPAEAFKQIS

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service