Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MTCT7955

MTC Bio Sample Transport Tubes

capacity 12 mL, round bottom

Synonym(s):

conical tube, reagent dish, sample storage tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121706
NACRES:
NE.25

material

clear
polypropylene
round bottom
screw cap

sterility

sterile

feature

graduation

packaging

pack of 1000 ea (10 x bags 100 ea)

manufacturer/tradename

MTC Bio T7955

capacity

12 mL

diam. × L

16 mm × 100 mm

Looking for similar products? Visit Product Comparison Guide

General description

16 x 100 mm screw cap test tubes are available in transparent polypropylene with a “skirted” self-standing bottom or in crystal clear polystyrene with a traditional round bottom. Each tube has easy to read graduations and a “frosted” white marking area (12x50mm). Packed 100 tubes (with attached caps) per sterile bag, 10 bags per unit.
Transport/ Culture Tube, 16 x1 00mm, PS, round bottom, with screw-cap, white printed graduations, 12mL, sterile, 10 bags of 100 tubes, 1000/cs

Features and Benefits

  • Leak-proof screw caps
  • White printed graduations
  • Sterile bags of 100 tubes
  • White marking spot
  • Self-standing / round bottom

Legal Information

MTC is a trademark of MTC Bio

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service