MilliporeSigma
MTCB1203
MTC Bio MultiBox for Western Blotting
4 compartments, L × W × D 32 mm (11/4 in.) × 52 mm (2 5/16 in.) × 28 mm (1 1/8 in.), for various gels
Synonym(s):
gel processing box, multi gel processing box, MTC Bio B1203, western blot apparatus, western blot chamber
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.15
UNSPSC Code:
41121800
Manufacturer/tradename:
MTC Bio B1203
Material:
polystyrene
Packaging:
pack of 6 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polystyrene
packaging
pack of 6 ea
manufacturer/tradename
MTC Bio B1203
L × W × D
32 mm (11/4 in.) × 52 mm (2 5/16 in.) × 28 mm (1 1/8 in.)
General description
MultiBox, 4 compartments, 32 x 52 x 28mm each (11/4 x 2 5/16 x 11/8 in.), for various gels, 6/pk
The MultiBox Western Blotting Box provides the ease and convenience of using one box to process multiple gels, allowing users to compare results side by side in real time.
Features and Benefits
  • A perfect box for every blot
  • Reduce antibody usage
  • Hinged lid minimizes evaporation
  • For halves, wholes, strips or minis
Legal Information
MTC is a trademark of MTC Bio
MultiBox is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
MTCB120304065270472113
MTCB1203-6EA04065271079212