Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

MBD0054

Sigma-Aldrich

Nucleic Acid Preservation buffer, microbial DNA free

Stabilizes and protects DNA and RNA

Synonym(s):

Stabilization buffer, NGS preservation buffer,microbiome preservation buffer, sample storage solution, nucleic acidstabilizer, nucleic acid protector, NAP buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.52

Quality Level

form

liquid

usage

Upon collection of the sample add 1-2 volumes of preservation buffer

shipped in

ambient

storage temp.

2-30°C

General description

Nucleic Acid Preservation buffer, microbial DNA free is a ready-made liquid, non-toxic tissue and cell storage reagent that stabilizes and protects cellular RNA and DNA in intact, unfrozen tissue and cell samples. Tested and found to be very effective for microbiome sample preservation. Nucleic Acid Preservation buffer eliminates the need to immediately process samples or to freeze samples for later processing.

Application

NucleicAcid Preservation buffer, microbial DNA free may be used as an effective preserving agent for complex samples at ambient temperature.

Features and Benefits

  • Ready-made solution
  • Preserves quality of DNA and RNA from fresh, unfrozen tissue and cell samples
  • Microbial DNA free for improved DNA analysis, especially important for low biomass samples
  • Immediately inhibits the activity of nucleases
  • Non-toxic, safe to transport
  • Stabilizes samples at room temperature for transport up to 10 days for DNA preservation and 6 days for RNA preservation
  • Effective for microbiome sample preservation

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service