MilliporeSigma
All Photos(4)
Key Documents
HS120679
Dual Purpose Scoops
size large, black, sterile; electron beam irradiated
About This Item
UNSPSC Code:
41122403
NACRES:
NB.71
material
black
polystyrene
size
large
sterility
sterile; electron beam irradiated
packaging
pack of 100 ea
manufacturer/tradename
Heathrow Scientific 120679
L × W × H
127 mm × 88.9 mm × 0.5 mm
General description
The sterile dual purpose scoop is a unique economical solution that combines the function of a scoop and a weigh boat into oneproduct. The weighing process has been streamlined, now just sample or retrieve the material in the scoop and place it directly on the scale. This eliminates the need for a weigh boat or weighing paper and saves valuable space in the lab. When finished the dual solution scoop is disposable.
Features and Benefits
  • Sterile
  • Made from anti-static polystyrene that is waterproof and reduces the chance of materials sticking to the scoop
  • Ships and stores flat and when ready to use, is easy to assemble by inserting the locking tab into the hole
  • Locking tab feature enables a scoop shape to help retain materials
  • Coated to stand up to powders, gels, and semi-solids
  • Broad tip design keeps the scoop stable while on the scale and enables materials to be easily scooped and dispensed
  • Disposable
Choose from one of the most recent versions:
Certificates of Analysis (COA)