Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE29401107

HiTrap® Capto Q

pack of 1 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NB.13

packaging

pack of 1 × 1 mL

manufacturer/tradename

Cytiva 29401107

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service