Skip to Content
MilliporeSigma
All Photos(1)

Documents

8305C

94090183, human thyroid, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

8305C, 94090183

biological source

human thyroid

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human thyroid anaplastic carcinoma, undifferentiated.

Cell Line Description

Established from undifferentiated thyroid carcinomas of a 67 year old female patient. Pathologically the carcinoma tissue contained residual well differentiated components suggesting well differentiated to undifferentiated carcinoma progression. Tumour suppresser genes p53, Rb, APC and MCC were analysed and sequence analysis confirmed a C:G to T:A transition at the first base of p53 gene codon 273. Loss of heterozygosity of tumour suppresser genes was not observed.

Application

As p53 mutations associate with undifferentiated carcinoma but not well differentiated carcinoma during multistep carcinogenesis of the thyroid, these cell lines should prove useful for research into the role of gene mutation in malignant transformation.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,12
D13S317: 9
D16S539: 10,11
D5S818: 10,13
D7S820: 8,10
THO1: 6,7
TPOX: 8
vWA: 14,16

Culture Medium

EMEM (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Doubling time is 43 hours. Saturation density at confluency is 5.7x10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service