Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COS-7 Cell Line

87021302, African green monkey kidney, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

COS-7 Cell Line, from African green monkey kidney, 87021302, SV40 transformed, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

African green monkey kidney

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Monkey African green kidney, SV40 transformed

Cell Line Description

Derived from CV-1 simian cells transformed by an origin-defective mutant of SV40. The cells support the growth of recombinant SV40 viruses. The cells should be handled at laboratory containment level 2.

Application

COS-7 Cell Line has been used to:
  • test the functional role of ankyrin B in ouabain/Na,K-ATPase/1,4,5-trisphosphate receptor (IP3R) signaling[1]
  • study the expression of green fluorescent protein-tagged constructs of neuropathy target esterase (NTE) and mutant proteins lacking the transmembrane, catalytic and regulatory domains[2]
  • study the distribution of peroxisomes[3]
Virus replication studies: SV40

Culture Medium

DMEM (D6546) + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442)..

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service