Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F9

85061803, mouse embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

F9, 85061803

biological source

mouse embryo

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse 129 embryo teratocarcinoma

Cell Line Description

Derived from testicular carcinoma. F9 undergo very limited differentiation under normal conditions but can be induced to differentiate into parietal endoderm in the presence of retinoic acid and dibutyryl cAMP.

Application

Differentiation studies

Culture Medium

DMEM + 2mM Glutamine + 10-15% Foetal Bovine Serum (FBS). Pre-soak flasks in 0.1% gelatin for over 2 hours and wash with PBS.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2, 37°C. Upon resuscitation, cells shed large quantities of debris, leave for 2 days before subculture. Please note: Culture v

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service