Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B65

85042305, rat nerve, Neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

B65, 85042305, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

rat nerve

packaging

tube of 5 μg 85042305-DNA-5UG
pkg of vial of cells 85042305-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat nervous tissue neuronal

Cell Line Description

Ethyl nitrosourea induced tumour cell line. Neuronal cells are useful for research into the distribution of neurotransmitter synthesis and brain specific antigens among nerve and glia. Cell morphology significantly affected when grown in serum free media.

Application

Neurobiological studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service