Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MM466

12070507, human skin (metastasis), Epithelial

Synonym(s):

MM 466

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

MM466, 12070507

biological source

human skin (metastasis)

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

receptors

Not specified

relevant disease(s)

metastasis

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Human melanoma metastatic site lymph node

Cell Line Description

mM466 is a human melanoma derived from a metastatic site (lymph node). Cell Bank Australia reference number CBA-1353.

DNA Profile

Not specified

Culture Medium

RPMI 1640 (R0883) (with 2 mM L-Glutamine (G7513) + 25mM HEPES) + 10% FBS.

Subculture Routine

Optimal split ratio 1:2-1:4 (seeding density 1.4 x 104 cells/cm2). Cells incubated at 37 °C with 5% CO2. Harvest the cells using 0.05% Trypsin/EDTA at 37 °C for 5 min. Medium change twice weekly. The cell line MM466 may take up to 72 hours to recover from thaw.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Peter Parsons - Queensland Institute of Medical Research, Australia

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service