Skip to Content
MilliporeSigma
All Photos(1)

Documents

CellVue® Claret Far Red Fluorescent Cell Linker Kit

Synonym(s):

Claret Cell Linker Kit, Far Red Fluorescent Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

storage temp.

room temp

Application

CellVue Claret kit fluoresces in the far red and may be used for general cell membrane labeling including in vitro cell labeling, in vitro lymphocyte proliferation studies, in vivo cytotoxicity assays, and also as a second color for monitoring cell-cell membrane transfer.

Biochem/physiol Actions

The CellVue Claret Fluorescent Cell Linker kit uses a proprietary membrane labeling technology to stably incorporate a far red fluorescent dye with long aliphatic tails (CellVue Claret) into lipid regions of the cell membrane.

Packaging

When using a 2 mL final staining volume containing a final concentration of 2 x 10-6 M of PKH26 dye:
  • the MINCLARET kit provides enough dye for up to 25 samples of 20 million cells each and enough diluent for up to 5 samples of 20 million cells each;
  • the MIDCLARET kit provides enough dye for up to 50 samples of 20 million cells each and enough diluent for up to 30 samples of 20 million cells each.
Users must determine the optimal dye and cell concentrations from their cell type(s) and experimental purposes.

Linkage

For additional technical details on Fluorescent Cell Linker Dyes including a comprehensive bibliography, please visit PKH and CellVue®; Claret Fluorescent Cell Linker Dyes.

Legal Information

Distributed for Phanos Technologies, Inc.
Patent Information
CellVue is a registered trademark of Phanos Technologies

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service