Documents
BAF187450012
Bel-Art® Switch-Grid® Test Tube Rack
Holds 40 x 16-20 mm tubes, green grid
Synonym(s):
tube holder
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
green grid
polyoxymethylene
feature
autoclavable
Holds 40 x 16-20 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18745-0012
L × W × H
31.8 cm × 11.1 cm × 6.7 cm
General description
1 RACK FRAME, 25 POSSIBLE TUBE SIZE COMBINATIONS
This innovative, split-frame rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Each rack frame includes a set of grids in one size. Grid sets in alternate sizes can be purchased as needed.
Features and Benefits
More compact and less expensive than purchasing multiple racks
Colored grid inserts correspond to a specific test tube size ranging from 10mm to 30mm in diameter
Three tier rack holds tubes securely in place
Rubber feet keep racks from sliding on benchtop, provide secure stacking while in storage
Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mild detergent, ethanol or isopropyl alcohol; autoclaved at 121°C (250°F).
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Switch-Grid is a registered trademark of SP Scienceware, a division of SP