Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV40640

Sigma-Aldrich

Anti-SYNCRIP (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Synaptotagmin binding, cytoplasmic RNA interacting protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

69 kDa

species reactivity

horse, mouse, bovine, rabbit, guinea pig, dog, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYNCRIP(10492)

Immunogen

Synthetic peptide directed towards the N terminal region of human SYNCRIP

Biochem/physiol Actions

Heterogenous nuclear ribonucleoprotein (hnRNP) implicated in mRNA processing mechanisms. SYNCRIP may be involved in translationally coupled mRNA turnover. It implicated with other RNA-binding proteins in the cytoplasmic deadenylation/translational and decay interplay of the FOS mRNA mediated by the major coding-region determinant of instability (mCRD) domain. It interacts in vitro preferentially with poly(A) and poly(U) RNA sequences.

Sequence

Synthetic peptide located within the following region: MATEHVNGNGTEEPMDTTSAVIHSENFQTLLDAGLPQKVAEKLDEIYVAG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service