Skip to Content
MilliporeSigma
All Photos(1)

Documents

AV39466

Sigma-Aldrich

Anti-NR2F2 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Nuclear receptor subfamily 2, group F, member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

dog, rabbit, human, guinea pig, sheep, horse, rat, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR2F2(7026)

General description

COUP transcription factor 2/nuclear receptor subfamily 2, group F, member 2 (COUP-TFII, NR2F2) is a nuclear receptor that regulates amygdale patterning via expression of neurophilin.COUP-TFII is a component of the glucagon-like peptide 1 (GLP-1) signaling cascade that stimulates neonatal β-cell number. COUP-TFII is required for GLP-1 activation of the β-catenin-dependent pathway.

Specificity

Anti-NR2F2 (AB1) polyclonal antibody reacts with chicken, bovine, pig, zebrafish, human, mouse, rat, and canine COUP transcription factor 2 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human NR2F2

Application

Anti-NR2F2 (AB1) polyclonal antibody is used to tag COUP transcription factor 2/nuclear receptor subfamily 2, group F, member 2 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of COUP transcription factor 2 in cell signaling pathways such as the glucagon-like peptide 1 (GLP-1) signaling cascade.

Biochem/physiol Actions

NR2F2, an orphan member of the nuclear hormone receptor superfamily, acts as a transcriptional repressor by antagonizing the functions of other nuclear hormone receptors and by actively silencing transcription. However, in certain contexts, NR2F2 stimulates transcription directly. NR2F2, MyoD and p300 interact in a competitive manner, and that increasing amounts of NR2F2 have the ability to reduce the interaction between myoD and p300 invitro. NR2F2 post-transcriptionally regulates myoD activity/function, and that crosstalk between orphan nuclear receptors and the myogenic bHLH proteins has functional consequences for differentiation.

Sequence

Synthetic peptide located within the following region: EYSCLKAIVLFTSDACGLSDVAHVESLQEKSQCALEEYVRSQYPNQPTRF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service