Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1749

Gold versus platinum thermocouple

NIST® SRM® 1749, certified thermometer for the range 0 °C to 1000 °C on the international temperature scale of 1990

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

packaging

pkg of each

manufacturer/tradename

NIST®

General description

This Standard Reference Material (SRM) is intended for use as a highly accurate secondary reference thermometer in the range of 0 °C to 1000 °C. For more information, please refer to the SDS and COA.

SRM 1749_cert

SRM 1749_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service