Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1089

Steels, set

NIST® SRM® 1089 (consists of SRMs 1095, 1096, 1097, 1098 and 1099)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 rods

manufacturer/tradename

NIST®

format

matrix material

General description

Other Notes

NIST SRM 1089 (consists of SRMs 1095, 1096, 1097, 1098 and 1099)
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Oxygen (O), Nitrogen (N), Hydrogen (H)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service