Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I0725000

Isopropyl hexadecanoate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Isopropyl palmitate, Isopropyl hexadecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOCH(CH3)2
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786567
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isopropyl palmitate

manufacturer/tradename

EDQM

refractive index

n20/D 1.438 (lit.)

mp

11-13 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC(C)C

InChI

1S/C19H38O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19(20)21-18(2)3/h18H,4-17H2,1-3H3

InChI key

XUGNVMKQXJXZCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isopropyl hexadecanoate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amnon C Sintov
International journal of pharmaceutics, 481(1-2), 97-103 (2015-02-07)
The objective of this study was to evaluate the transdermal delivery potential of a new curcumin-containing microemulsion system. Three series of experiments were carried out to comprehend the system characteristics: (a) examining the influence of water content on curcumin permeation
Rodrigo Molina Martins et al.
Journal of microencapsulation, 31(7), 644-653 (2014-04-29)
Solid-lipid microparticles loaded with high amounts of the sunscreen UV filter benzophenone-3 were prepared by spray congealing with the objective of decreasing its skin penetration and evaluate whether the sunscreen's photoprotection were impaired by the microencapsulation process. The microparticles were
Amnon C Sintov et al.
International journal of pharmaceutics, 471(1-2), 516-524 (2014-05-29)
The aim of this study was to explore the transdermal delivery potential of a new caffeine-containing microemulsion system. The skin permeability of caffeine (CAF) was measured in vitro using skin excised from three different animal species: rat, rabbit and pig.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service