Skip to Content
MilliporeSigma
All Photos(1)

Documents

BCR547

Acetaldehyde-2,4-dinitrophenylhydrazone

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
224.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

mp

165-168 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

C\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C8H8N4O4/c1-2-9-10-7-4-3-6(11(13)14)5-8(7)12(15)16/h2-5,10H,1H3/b9-2+

InChI key

ONBOQRNOMHHDFB-XNWCZRBMSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR547

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service