Skip to Content
MilliporeSigma
All Photos(4)

Documents

910430

Sigma-Aldrich

Grubbs Catalyst® M220

Umicore

Synonym(s):

cis-[1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(3-phenyl-1H-inden-1-ylidene)(triisopropylphosphite)ruthenium(II), Grubbs Catalyst® M22

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H57N2Cl2O3PRu
CAS Number:
Molecular Weight:
876.90
UNSPSC Code:
12352103
NACRES:
NA.22

form

powder

mp

>300 °C

storage temp.

2-8°C

Application

Umicore Grubbs Catalyst M220 is a homogeneous catalyst useful for cross metathesis, enyne metathesis, ring-closing metathesis, and self metathesis.

Learn more about our metathesis catalysts

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service