Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

229121

Sigma-Aldrich

Solvent Blue 59

Dye content 98 %

Synonym(s):

1,4-Bis(ethylamino)-9,10-anthraquinone, Atlasol Blue 2N, Sudan Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O2
CAS Number:
Molecular Weight:
294.35
Colour Index Number:
61552
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

≥97.5% (HPLC)

form

powder

composition

Dye content, 98%

mp

215-217 °C (lit.)

λmax

595 nm
640 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCNc1ccc(NCC)c2C(=O)c3ccccc3C(=O)c12

InChI

1S/C18H18N2O2/c1-3-19-13-9-10-14(20-4-2)16-15(13)17(21)11-7-5-6-8-12(11)18(16)22/h5-10,19-20H,3-4H2,1-2H3

InChI key

JUUJTYPMICHIEM-UHFFFAOYSA-N

General description

Solvent Blue 59 is an anthraquinone dye, also known as 1,4 Bis-(ethylamino)-anthraquinone.

Application

Solvent Blue 59 has been utilized in polycarbonate and thermoplastic polyesters due to its efficient heat stability.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Tanigawa et al.
Lymphology, 14(4), 149-154 (1981-12-01)
Sudan Black B and Sudan Blue can partly enter the lymphatics of the small intestine from the lumen with long chain fatty acids. By use of a fat emulsion saturated with them small intestinal and mesenteric lymphatics were clearly delineated.
Jawwad A Qureshi et al.
Journal of economic entomology, 97(3), 836-845 (2004-07-29)
Mark-release-recapture experiments to study insect dispersal require the release of marked insects that can be easily identified among feral conspecifics. Oil-soluble dyes have been used successfully to mark various insect species. Two oil-soluble dyes, Sudan Red 7B (C.I. 26050) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service