Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RW0309000

Millipore

MCE Membrane Filter, 0.2 μm Pore Size

Millipore, filter diam. 90 mm, hydrophilic

Synonym(s):

Reinforced Membrane Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

mixed cellulose esters (MCE) membrane
plain filter
web polyester membrane
white filter

Quality Level

description

90 mm diameter, mixed cellulose esters, 0.2 µm pore size, polyester web

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Millipore

parameter

50 mL/min-cm2 water flow rate
70 °C max. temp.

diam.

90 mm

filter diam.

90 mm

thickness

200 μm

pore size

0.2 μm pore size
75 % porosity

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Application

Prefiltration before 0.22 μm membrane filtration

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service