Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABT179

Sigma-Aldrich

Anti-Osterix (SP7) Antibody, clone 6B12.1

clone 6B12.1, from mouse

Synonym(s):

Transcription factor Sp7, Zinc finger protein osterix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6B12.1, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SP7(121340)

General description

OSTERIX, or SP7, is a zinc finger-containing, osteoblast-specific transcription factor that is distinctly expressed in all developing bones, and is a key requirement for the differentiation of pre-osteoblasts into fully functional osteoblasts. While research on the actual functional mechanisms of OSTERIX is still on-going, current findings indicate that OSTERIX expression is induced by BMP-2 and Dlx5, and operates downstream of RUNX2, supporting its role in mesenchymal stem cell differentiation. Further studies also indicate that the OSTERIX transcription factor may also play a role in negatively regulating the Wnt/β-catenin signaling pathways and the proliferation of osteroblast cell types during embryonic osteogenesis.

Immunogen

GST-tagged recombinant protein corresponding to human Osterix.

Application

Detect the Osterix (SP7) protein using this Anti-Osterix (SP7), clone 6B12.1 validated for use in WB & IHC.
Immunohistochemistry Analysis: A 1:200 dilution from a representative lot detected Isocitrate dehydrogenase in rat tooth tissue. Nuclear staining may be observed in epithelial cells in some tissues.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blotting in SW1353 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Osterix in 10 µg of SW1353 cell lysate.

Target description

~45 kDa observed. An uncharacterized band may appear at ~160 kDa in some lysates.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
SW1353 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service