Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABS1214

Sigma-Aldrich

Anti-PEG (methoxy group) Antibody, clone RM105

clone RM105, from rabbit, purified by affinity chromatography

Synonym(s):

PEG, pegylation, methoxy group, Polyethylene glycol, polyethylene oxide, PEO, polyoxyethylene, POE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM105, monoclonal

purified by

affinity chromatography

species reactivity (predicted by homology)

all

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

General description

Polyethylene glycol (PEG) is a polyether compound with many applications from industrial manufacturing to medicine. PEG is also known as polyethylene oxide (PEO) or polyoxyethylene (POE), depending on its molecular weight. PEG is a family of long chain polymers attached to a glycerine backbone. Some of the properties of PEG are nonionic, nontoxic, biocompatible, strongly hydrophilic polymer, which has a large exclusion volume in aqueous solution. PEGylation is the act of covalently coupling a PEG structure to another larger molecule, for example, a therapeutic protein, which is then referred to as a PEGylated protein. The covalent attachment of PEG (PEGylation) is commonly used to modify a variety of proteins and drugs. Some of the therapeutical benefits of PEGylated molecules include increased bioavailability, increased blood circulation of the drug, optimized pharmacokinetics, decreased immunogenicity, and decreased frequency of administration. Additional benefits of PEGylated molecules include decelerated renal excretion, improved stability towards proteolysis, increased solubility of the biopharmaceutical in aqueous solutions, increased hydrodynamic radius, reduced immunogenicity and proteolytic cleavage. This antibody reacts to the terminal methoxy group of Polyethylene glycol.

Immunogen

recombinant protein corresponding to All PEG (methoxy group).

Application

Anti-PEG (methoxy group), clone RM105 is an antibody against PEG (methoxy group) for use in WB, IH, ELISA.
Immunohistochemistry Analysis: 0.5 μg/mL of this antibody detected PEG (methoxy)-BSA in injected mouse liver and kidney
ELISA Analysis: Multiple concentrations of this antibody was used to detect three different BSA-PEG conjugates (5K, 40K and 40K branched)
Research Category
Signaling
Research Sub Category
Epitope Tags

Quality

Evaluated by Western Blotting on recombinant methoxyPEG conjugated BSA

Western Blotting Analysis: A 1:1;000 dilution of this antibody detected PEG (methoxy group) on conjugated recombinant BSA

Target description

BSA is 66 kD, methoxyPEG is 5 kD. Additional bands above 66 kD is due to different levels of conjugation and dimerization.

Physical form

Protein A purified
Purified rabbit monoclonal IgG in PBS buffer containing 1% BSA and 0.09% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng-ying Huang et al.
European journal of cancer (Oxford, England : 1990), 48(14), 2260-2269 (2012-01-20)
Cytochalasin D targets actin and is ubiquitous in eukaryotic cells. When cytochalasin D is used as a cytotoxic agent in cancer therapy, it causes significant side effects. To prevent this, cytochalasin D can be encapsulated in polyethylene liposomes. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service