Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN2284

Sigma-Aldrich

Anti-Delta-like protein 1 Antibody, clone PGPM-1F9

clone PGPM-1F9, from rat

Synonym(s):

Drosophila Delta homolog 1, Delta1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PGPM-1F9, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Dll1(13388)

Related Categories

General description

Delta-like protein 1 (UniProt: Q61483; also known as Drosophila Delta homolog 1, Delta1, DLL1) is encoded by the Dll1 gene (Gene ID: 13388) in murine species. DLL1 is a single-pass transmembrane ligand protein of NOTCH1, NOTCH2 and NOTCH3 receptors that binds the extracellular domain (ECD) of Notch receptor in a cis and trans fashion manner. Following transinteraction, ligand cells produce mechanical force that promotes Notch receptor extracellular domain (NECD) transendocytosis and triggers Notch signaling. Delta1 is required for embryonic development and maintenance of adult stem cells in many different tissues. DLL1-induced Notch signaling plays a role in the regulation of cell lineage, cell specification, cell patterning, and morphogenesis through effects on differentiation and proliferation. DLL1 is reported to play a role in regulating neuronal differentiation of neural precursor cells via cell-cell interaction. DLL1 is reported to be co-expressed with DLL3 in the presomtic mesoderm of mouse embryo, however despite their co-expression, mutations in these proteins cause strikingly different effects. Heterozygous DLL1 mutant mice are lighter and smaller, with altered fat to lean ratio and display increased blood pressure and a slight bradycardia. They also exhibit reduced cholesterol and triglyceride levels in blood.

Specificity

Clone PGPM-1F9 specifically detects murine Delta Like protein 1. It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal half of murine Delta-like protein 1.

Application

Anti-Delta-like protein 1 antibody, clone PGPM-1F9, Cat. No. MABN2284, is a highly specific rat monoclonal antibody that targets Delta-like protein 1 and has been tested in Flow Cytometry, Immunohistochemistry, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected Delta-like protein 1 in Immunohistochemistry applications (Bone, R.A., et. al. (2014). Development. 141(24):4806-16).

Flow Cytometry Analysis: 1 µg from a representative lot detected Delta-like protein 1 in one million DLL1-flag A7A1/CHO cells expressing DLL1.

Western Blotting Analysis: A representative lot detected Delta-like protein 1 in Western Blotting applications (Geffers, I., et. al. (2007). J Cell Biol. 178(3):465-76).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in DLL1-FLAG/CHO cell lysates.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Delta-like protein 1 in 10 µg of lysate from CHO cells stably expressing Flag-tagged Delta-like Protein 1.


Target description

~78 kDa observed; 78.45 calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brittany M Edens et al.
Cell reports, 28(4), 845-854 (2019-07-25)
N6-methyladenosine (m6A) modification of mRNA is emerging as a vital mechanism regulating RNA function. Here, we show that fragile X mental retardation protein (FMRP) reads m6A to promote nuclear export of methylated mRNA targets during neural differentiation. Fmr1 knockout (KO)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service