Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABF3178

Sigma-Aldrich

Anti-IL-22 Antibody, clone 9F2.2.4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies, primary antibodies

clone

9F2.2.4, monoclonal

mol wt

calculated mol wt 20.01 kDa
observed mol wt ~17 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... IL22(50616)

General description

Interleukin-22 (UniProt: Q9GZX6; also known as IL-22, Cytokine Zcyto18, IL-10-related T-cell-derived-inducible factor, IL-TIF) is encoded by the IL22 (also known as ILTIF, ZCYTO18, UNQ3099/PRO10096) gene (Gene ID: 50616) in human. IL-22 is an -helical cytokine that contributes to the inflammatory response in vivo. It is a secretory protein that is synthesized with a signal peptide (aa 1-33), which is subsequently cleaved off to produce the mature protein. It is immediately produced to initiate immune response against several tissue impairments. It is considered as a fundamental mediator in inflammation, mucous production, protective role against pathogens, wound healing, and tissue regeneration. It is produced by several populations of immune cells, including macrophages, β T cells classes Th1, Th22, and Th17 cells at the site of inflammation. It is shown to up-regulate a number of mucus associated genes, such as MUC1, MUC3, MUC10, and MUC13 in colonic and tracheal epithelial cells. Along with IL-17A, it also produces pulmonary mucus as a control against Gram negative pathogens. Its biological activity is initiated by binding to a cell-surface complex composed of IL-22R1 and IL-10R2 receptor chains and further regulated by interactions with a soluble binding protein (IL-22BP). IL-22 is shown to have a very high affinity for binding to IL-22BP, which acts as a natural antagonist of IL-22. IL-22BP binding occurs at specific sites on IL-22 that are also utilized by IL-22R1. (Ref.: Arshad, T., et al. (2020). Front. Immunol. 11; 2148).

Specificity

Clone 9F2.2.4 is a mouse monoclonal antibody that detects human IL-22.

Immunogen

A full-length mature human IL-22 (without the signal peptide).

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant human IL-22.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant human IL-22.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected IL-22 in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IL-22, clone 9F2.2.4, Cat. No. MABF3178, is a mouse monoclonal antibody that detects human IL-22 and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service