Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABF2794

Sigma-Aldrich

Anti-TOX Antibody, clone NAN448B

Synonym(s):

Thymocyte selection-associated high mobility group box protein TOX;Thymus high mobility group box protein TOX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

NAN448B, monoclonal

mol wt

calculated mol wt 57.51 kDa
observed mol wt ~57 and 61 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TOX(9760)

General description

Thymocyte selection-associated high mobility group box protein TOX (UniProt: O94900; also known as Thymus high mobility group box protein TOX) is encoded by the TOX (also known as KIAA0808) gene (Gene ID: 9760) in human. TOX is a member of an evolutionarily conserved DNA-binding protein family that plays multiple roles in the development of the adaptive immune system. It acts as a developmental checkpoint and regulates thymocyte positive selection toward T cell lineage commitment. It is required for the development of various T cell subsets, including CD4+ helper T-cells, CD8+ cytotoxic T-cells, regulatory T-cells, and CD1D-dependent natural killer T (NKT) cells. Its nuclear localization signal lies in amino acids 237-256 and the DNA-binding region (HMG box) resides in amino acids 161-329. TOX serves as a DNA-binding factor that regulates transcription by modifying local chromatin structure and modulating the formation of multi-protein complexes. It binds to GC-rich DNA sequences in the proximity of transcription start sites to alter chromatin structure and modify access of transcription factors to DNA. It also plays a major role in neural stem cell commitment and corticogenesis. During cortical development, it controls the neural stem cell pool by inhibiting the switch from proliferative to differentiating progenitors. It also promotes neurite outgrowth in newborn neurons migrating to reach the cortical plate. In T-cell lymphomas high TOX expression has been described in all the Mycosis fungoides (MF). A majority of precursor B/T lymphoblastic, follicular and diffuse large B-cell lymphomas, nodular lymphocyte-predominant Hodgkin lymphomas and angioimmunoblastic T-cell lymphomas strongly expressed the TOX protein. (Ref.: Maestre, L., et al. (2020). PLoS ONE 15(2); e0229743; Yun, S., et al. (2011). Immunol. Lett. 136(1); 29-36).

Specificity

Clone NAN448B is a rat monoclonal antibody that detects human Thymocyte selection-associated high mobility group box protein TOX. It targets an epitope within the N-terminal half

Immunogen

His-tagged recombinant fragment corresponding to the first 250 amino acids form human Thymocyte selection-associated high mobility group box protein TOX (TOX).

Application

Quality Control Testing

Evaluated by Western Blotting in MOLT-4 cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TOX in MOLT-4 cell lysates.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected TOX in Immunocytochemistry applications (Maestre, L., et al. (2020). PLoS One. 15(2):e0229743).

Immunohistochemistry Applications: A representative lot detected TOX in Immunohistochemistry applications (Maestre, L., et al. (2020). PLoS One. 15(2):e0229743).

Immunofluorescence Analysis: A representative lot detected TOX in Immunofluorescence applications (Maestre, L., et al. (2020). PLoS One. 15(2):e0229743).

Western Blotting Analysis: A representative lot detected TOX in Western Blotting applications (Maestre, L., et al. (2020). PLoS One. 15(2):e0229743).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TOX, clone NAN448B, Cat. No. MABF2794, is a rat monoclonal antibody that detects Thymus high mobility group box protein TOX and is tested for use in Immunocytochemistry, Immunofluorescence, Immunohistochemistry, and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service