Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2323

Sigma-Aldrich

Anti-HLA-A02:01 EBV-EBNA1 Antibody, clone MAB13

Synonym(s):

Epstein-Barr nuclear antigen 1;EBNA-1;EBV nuclear antigen 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MAB13 (also referenced as E1), monoclonal

mol wt

calculated mol wt 56.43 kDa

purified by

using protein G

species reactivity

human, mouse

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... EBNA1(3783774)

General description

Epstein-Barr nuclear antigen 1 (UniProt: P03211; also known as EBNA-1, EBV nuclear antigen 1) is encoded by the EBNA1 (also known as BKRF1) gene (Gene ID: 3783709) in Epstein-Barr virus (EBV). EBV is a gamma-herpesvirus, a causative agent of infectious mononucleosis. Although the majority of virus infected individuals remain healthy, EBV has been implicated in the pathogenesis of African Burkitt s lymphoma and nasopharyngeal carcinoma. EBNA1, a multi-functional protein, plays an important role in maintaining EBV in the host cell and has been implicated in host cell immortalization. It plays an essential role in replication and partitioning of viral genomic DNA during latent viral infection. During this phase, the circular double-stranded viral DNA undergoes replication once per cell cycle and is efficiently partitioned to the daughter cells. EBNA1 activates the initiation of viral DNA replication through binding to specific sites in the viral latent origin of replication, oriP. It is also reported to direct the segregation of viral episomes by mediating their attachment to host cell metaphase chromosomes and is known to activate the transcription of several viral latency genes. By counteracting the stabilization of host p53, it also reduces apoptosis and increases host cell survival. EBNA1 is also a risk factor in a variety of malignancies and is the only viral protein found in all EBV-related malignancies. Its C-terminal region contains a dimerization domain and a DNA binding domain (aa 459-607). Although this domain is essential for its replication function of EBNA1, it is not sufficient for this activity and the N-terminal half of EBNA1 is also required.

Specificity

Clone MAB13 (E1) is a mouse monoclonal antibody that detects EBNA1. It targets an epitope within 9 amino acids from the C-terminal region.

Immunogen

Membrane free HLA-A201 with EBV (strain B95-8) EBNA1 (FMVFLQTHI) 562-570 recombinant protein.

Application

Quality Control Testing

Evaluated by Flow Cytometry in T2 cells pulsed with EBNA1 peptide (562-570).

Flow Cytometry Analysis (FC): 1 μg of this antibody detected HLA-A2-EBV EBNA1 in one million T2 cells pulsed with EBNA1 peptide (562-570).

Tested Applications

Immunofluorescence Analysis: A representative lot detected HLA-A02:01 EBV-EBNA1 in Immunofluorescence applications (Lai, J., et al. (2016). Blood. 128(10):1396-407; Sim, A.C.N., et al. (2013). Sci Rep. 3:3232).

Immunohistochemistry Applications: A representative lot detected HLA-A02:01 EBV-EBNA1 in Immunohistochemistry applications (Sim, A.C.N., et al. (2013). Sci Rep. 3:3232).

Western Blotting Analysis: A representative lot detected HLA-A02:01 EBV-EBNA1 in Western Blotting applications (Sim, A.C.N., et al. (2013). Sci Rep. 3:3232).

Flow Cytometry Analysis: A representative lot detected HLA-A02:01 EBV-EBNA1 in Flow Cytometry applications (Lai, J., et al. (2016). Blood. 128(10):1396-407; Lai, J., et al. (2017). Sci Rep. 7(1):9923).

Immunocytochemistry Analysis: A representative lot detected HLA-A02:01 EBV-EBNA1 in Immunocytochemistry applications (Lai, J., et al. (2016). Blood. 128(10):1396-407; Lai, J., et al. (2017). Sci Rep. 7(1):9923).

Surface plasmon resonance: A representative lot detected HLA-A02:01 EBV-EBNA1 in Surface plasmon resonance applications (Sim, A.C.N., et al. (2013). Sci Rep. 3:3232).

ELISA Analysis: A representative lot detected HLA-A02:01 EBV-EBNA1 in ELISA applications (Lai, J., et al. (2016). Blood. 128(10):1396-407; Lai, J., et al. (2017). Sci Rep. 7(1):9923).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-HLA-A02:01 EBV-EBNA1, clone MAB13, Cat. No. MABF2323, is a mouse monoclonal antibody that detects EBNA1 and is used in ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Surface Plasmon Resonance & Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service