Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABC1613

Sigma-Aldrich

Anti-MUC1 Antibody, clone HMFG2

clone HMFG2, from mouse

Synonym(s):

Mucin-1, Breast carcinoma-associated antigen DF3, Cancer antigen 15-3, CA 15-3, Carcinoma-associated mucin, Episialin, H23AG, Krebs von den Lungen-6, KL-6, PEMT, Peanut-reactive urinary mucin, PUM, Polymorphic epithelial mucin, PEM, Tumor-associated epit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HMFG2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1λ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MUC1(4582)

General description

Mucin-1 (UniProt: P15941; MUC-1, Breast carcinoma-associated antigen DF3, Cancer antigen 15-3, CA 15-3, Carcinoma-associated mucin, Episialin, H23AG, Krebs von den Lungen-6, KL-6, PEMT, Peanut-reactive urinary mucin, PUM, Polymorphic epithelial mucin, PEM, Tumor-associated epithelial membrane antigen, EMA, Tumor-associated mucin, CD227) is encoded by the MUC1 (also known as PUM) gene (gene ID: 4582) in human. The MUC-1 alpha subunit has cell adhesive properties and functions as protective layer on epithelial cells against bacteria and enzymes. It can act as both cell adhesion and an anti-adhesion protein. It forms a tight, non-covalent heterodimeric complex with the proteolytically-released beta-subunit. The beta subunit contains a C-terminal domain that is involved in cell signaling via phosphorylations and protein-protein interactions. It modulates signaling in ERK, SRC and NF-kappa-B pathways. In activated T-cells, it impacts directly or indirectly the Ras/MAPK pathway. MUC-1 also is involved in tumor progression and transcription through regulation of Tp53. MUC-1 is expressed in epithelial cells, breast, uterus, activated and unactivated T-cells. It is shown to be over-expressed in epithelial tumors, such as breast or ovarian cancer. MUC-1 is exclusively located in the apical domain of the plasma membrane of highly polarized epithelial cells. After endocytosis, it is internalized and recycled to the cell membrane. MUC-1 is highly glycosylated (N- and O-linked carbohydrates and sialic acid) and is O-glycosylated to a varying degree on serine and threonine residues within each tandem repeat, ranging from mono- to penta-glycosylation. The average density ranges from about 50% in human milk to over 90% in T47D breast cancer cells. Its interaction, via the tandem repeat region, with domain 1 of ICAM1 is implicated in cell migration and metastases. MUC1 serves as a serological clinical marker of breast cancer to monitor response to breast cancer treatment and disease recurrence. Lower levels over time may be indicative of a positive response to treatment.

Specificity

Clone HMFG2 immunostains human breast cancer tissue. It can react with the unglycosylated peptide as well as several glycoforms of MUC1.

Immunogen

Delipidated milk fat globule extract followed 3 weeks later by epithelial cells cultured from milk (for 2 weeks).

Application

Detect Mucin-1 using this mouse monoclonal Anti-MUC1, clone HMFG2, Cat. No. MABC1613 that has been tested for use in Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Functional studies, and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected MUC1 in human lung and human pancreas tissue.

Western Blotting Analysis: A representative lot detected MUC1 in Western Blotting applications (Kaur, S., et. al. (2014). PLoS One. 9(3):e92742; Schroeder, J.A., et. al. (2001). J Biol Chem. 276(16):13057-64).

Affects Function: A representative lot detected MUC1 in Affects Function applications (Schettini, J., et. al. (2012). Cancer Immunol Immunother. 61(11):2055-65).

Immunoprecipitation Analysis: A representative lot detected MUC1 in Immunoprecipitation applications in (Schroeder, J.A., et. al. (2001). J Biol Chem. 276(16):13057-64).

Immunohistochemistry Analysis: A representative lot detected MUC1 in Immunohistochemistry applications in (Sakurai, J., et. al. (2007). Eur J Histochem. 51(2):95-102).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry in human breast cancer tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected MUC1 in human breast cancer tissue.

Target description

122.10 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service