Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43845

Sigma-Aldrich

Selenium disulfide

for synthesis

Synonym(s):

Selenium disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S2Se
CAS Number:
Molecular Weight:
143.09
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-303-8
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

solid

potency

138 mg/kg LD50, oral (Rat)

storage temp.

2-30°C

SMILES string

[Se](=S)=S

InChI

1S/S2Se/c1-3-2

InChI key

JNMWHTHYDQTDQZ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service