Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.07339

Sigma-Aldrich

Platinum/charcoal activated

10% Pt, hydrogenation catalyst for synthesis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.22

product name

Platinum/charcoal activated (10% Pt), hydrogenation catalyst for synthesis

Quality Level

autoignition temp.

600 °C

pH

2-10 (20 °C, 50 g/L in H2O)

bulk density

250 kg/m3

storage temp.

2-30°C

Analysis Note

Assay (of Pt): 9.0 - 11.0 %
Suitable for hydrogenation catalyst: passes test
Identity ( RFA): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service