Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

5.38765

Sigma-Aldrich

PDI Inhibitor IV, LOC14

Synonym(s):

PDI Inhibitor IV, LOC14, 2-((4-(Cyclopropanecarbonyl)piperazin-1-yl)methyl)benzo[d]isothiazol-3(2H)-one, 2-((4-(Cyclopropylcarbonyl)-1-piperazinyl)methyl)-1,2-benzothiazol-3(2H)-one, Lead Optimized Compound 14, Protein Disulfide Isomerase Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O2S
Molecular Weight:
317.41
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, blood brain barrier permeable, orally bioavailable, non-toxic benzothiazolone derived compound that acts a potent, reversible and non-covalent inhibitor of protein disulfide isomerase (PDI) reductase activity (Kd = 61.7 nM for PDIa). Shown to bind to PDIa stoichiometrically (1:1), adjacent to the active site, and enforce PDI to adopt an oxidized conformation. Dose-dependently rescues both PC12 cells (EC50 = 500 nM) and medium spiny neurons from mutant huntingtin neurotoxicity. Exhibits favorable microsomal (k = 0.0016, t1/2 = 438.7 min, CLint = 0.17 ml/min/g mouse liver) and blood plasma stability (k = 0.0049, t1/2 = 144 min and 49.9% remains after 2 h incubation) and is well tolerated in mice (20 mg/kg, p.o.).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDI
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only anhydrous DMSO for reconstitution.

Other Notes

Kaplan, A., et al. 2015. Proc. Natl. Acad. Sci. USA112, In press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service