Skip to Content
MilliporeSigma
All Photos(1)

Documents

529583-M

Sigma-Aldrich

Prekallikrein, Human Plasma

Synonym(s):

Fletcher Factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

specific activity

22-28 U/mg (clotting assay)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

Native prekallikrein from human plasma. Single-chain glycoprotein that participates in the early phase of blood coagulation, kinin formation, and fibrinolysis.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 4 mM NaOAc-HCl, pH 5.3.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Shariat-Madar, Z., et al. 1999. J. Biol. Chem.274, 7137.
Rojkjaer, R., et al. 1998. Thromb. Haemost.80, 74.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service