Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444218

Sigma-Aldrich

MMP-3 Inhibitor I

The MMP-3 Inhibitor I controls the biological activity of MMP-3. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-3 Inhibitor I, Ac-RCGVPD-NH₂, Stromelysin-1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46N10O9S
Molecular Weight:
686.78
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Inhibits matrix metalloproteinase-3 (stromelysin) (IC50 = 5 µM). The inhibitor is based on a segment from the conserved region of the NH2-terminal propeptide domain.
Inhibits the matrix metalloproteinase MMP-3 (IC50 = 5 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMP-3
Product does not compete with ATP.
Reversible: no
Target IC50: 5 µM against MMP-3

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Arg-Cys-Gly-Val-Pro-Asp-NH₂

Physical form

Supplied as a trifluroacetate salt.

Reconstitution

Following recosntitution, aliquot and freeze (-20°C). Stock solutions are stable for 1-2 weeks at -20°C.

Other Notes

Fotouhi, N., et al. 1994. J. Biol. Chem. 269, 30227.
Hanglow, A.C., et al. 1993. Agents Actions 39, C148.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service