Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341501

Sigma-Aldrich

FeTMPyP

Catalyzes the isomerization of peroxynitrite (ONOO⁻) to nitrate (NO3⁻) under physiological conditions and at realistic dose-derived concentrations (kcat = 2.2 x 10⁶M⁻¹s⁻¹).

Synonym(s):

FeTMPyP, 5,10,15,20-Tetrakis(N-methyl-4ʹ-pyridyl)porphinato Iron (III) Chloride, Fe IIITMPyP(Cl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H36Cl5FeN8
Molecular Weight:
909.92
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

brown

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Catalyzes the isomerization of peroxynitrite (ONOO) to nitrate (NO3) under physiological conditions and at realistic dose-derived concentrations (Kcat = 2.2 x 106 M-1s-1). Effectively prevents the oxidation of lipid components by peroxynitrite. Also catalytically decomposes superoxide radicals (O2·). Irradiation of the aqueous solution at 335 nm releases hydroxyl (OH·) radicals. FeTMPyP can also serve as a potential for drug detoxification of reactive oxygen species (ROS).

Biochem/physiol Actions

Cell permeable: no
Kcat = 2.2 x 106 M-1s-1 in catalyzing the isomerization of peroxynitrite (ONOO-) to nitrate (NO3-) under physiological conditions
Primary Target
Peroxynitrite (ONOO-)
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Lee, J., et al. 1998. J. Am. Chem. Soc. 120, 7493.
Hunt, J.A., et al. 1997. Chem. Biol. 4, 845.
Marla, S.S., et al. 1997. Proc. Natl. Acad. Sci. USA 94, 14243.
Stern, M.K., et al. 1996. J. Am. Chem. Soc. 118, 8735.
Faraggi, M., et al. 1984. Int. J. Radiat. Biol. Relat. Stud. Phys. Chem. Med.46, 703.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service