Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.66506

Millipore

Tryptic Soy Agar

with Lecithin and Tween®, ICR Settle plate, irradiated, triple packed, for environmental monitoring (Isolator and Clean room)

Synonym(s):

TSA, Soybean Casein Digest Agar, ICR Settle plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

2D bar code
ready-to-use

packaging

pkg of (20/120 Plates per box)

D

90 mm

pH

(7.3 (in H2O))

application(s)

air monitoring
microbiology
pharmaceutical

storage temp.

15-25°C

suitability

bacteria
fungi

General description

Tryptic Soy Agar LT - ICR settle plate is designed for the determination of the total microbial aerobic count in Clean rooms and Isolators. Settle plates are used for air and personnel monitoring. Tryptic soy agar is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. The medium is supplemented with Lecithin, Polysorbate 80 (Tween®) and pyruvate to inactivate potential residues of disinfectants.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic count.

Packaging

1665060020-CN: 20 Plates per box
1665060120-CN: 120 Plates per box

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service