Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.19233

Supelco

Platinum black 98+

for analysis 98+ EMSURE®

Synonym(s):

Platinum black 98+

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
EC Index Number:
231-116-1
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

≥98.0% (calculated on calcined substance)

impurities

≤10 ppm Al (Aluminium)
≤10 ppm Cd (Cadmium)
≤10 ppm Co (Cobalt)
≤10 ppm Cr (Chromium)
≤10 ppm Mn (Manganese)
≤10 ppm Ni (Nickel)
≤100 ppm Au (Gold)
≤100 ppm Ca (Calcium)
≤100 ppm Fe (Iron)
≤100 ppm Hg (Mercury)
≤100 ppm Ir (Iridium)
≤100 ppm Os (Osmium)
≤100 ppm Pd (Palladium)
≤100 ppm Rh (Rhodium)
≤100 ppm Ru (Ruthenium)
≤200 ppm Zn (Zinc)
≤50 ppm Ag (Silver)
≤50 ppm Cu (Copper)
≤50 ppm K (Potassium)
≤50 ppm Mg (Magnesium)
≤50 ppm Pb (Lead)

mp

1770 °C

density

21.45 g/cm3

storage temp.

15-25°C

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Analysis Note

Identity (elemental analysis): passes test
Assay (calculated on calcined substance): ≥ 98.0 %
Ag (Silver): ≤ 50 ppm
Al (Aluminium): ≤ 10 ppm
Au (Gold): ≤ 100 ppm
Ca (Calcium): ≤ 100 ppm
Cd (Cadmium): ≤ 10 ppm
Co (Cobalt): ≤ 10 ppm
Cr (Chromium): ≤ 10 ppm
Cu (Copper): ≤ 50 ppm
Fe (Iron): ≤ 100 ppm
Hg (Mercury): ≤ 100 ppm
Ir (Iridium): ≤ 100 ppm
K (Potassium): ≤ 50 ppm
Mg (Magnesium): ≤ 50 ppm
Mn (Manganese): ≤ 10 ppm
Ni (Nickel): ≤ 10 ppm
Os (Osmium): ≤ 100 ppm
Pb (Lead): ≤ 50 ppm
Pd (Palladium): ≤ 100 ppm
Rh (Rhodium): ≤ 100 ppm
Ru (Ruthenium): ≤ 100 ppm
Zn (Zinc): ≤ 200 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service