Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.18463

Supelco

Nitrite Test Refill pack

for use with MCOLORTEST®

Synonym(s):

Nitrite Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

feature

refill

Quality Level

specific analyte(s)

nitrite

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

This refill pack contains all the reagents for the MQuant® test kits 1.14774, 1.14424, 1.14408 for the colorimetric evaluation of nitrite.

Method
In acidic solution nitrite ions react with sulfanilic acid to form a diazonium salt, which in turn reacts with N-(1-naphthyl)ethylenediamine dihydrochloride to form a red-violet azo dye. The nitrite concentration is measured semiquantitatively by visual colorimetric evaluation.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service