Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

1.11860

Supelco

Blank Test Strips, colorimetric

greener alternative

MQuant®

Synonym(s):

Blank Strips chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Blank Strips, colorimetric, MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

analyte chemical class(es)

free fatty acids, sugars

specific analyte(s)

aluminium, ammonium, ammonium, arsenic, ascorbic acid, calcium, calcium, chloride, chlorine, chromate, copper, cyanide, fluoride, formaldehyde, hydrogen peroxide, iron, lead, magnesium, manganese, molybdenum, nickel, nitrate, nitrite, peracetic acid, peroxidase, phosphate, potassium, sulfate, sulfide, sulfite, tin, zinc

reaction suitability

reagent type: oxidant

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

The MQuant® blank test strips incorporate a paperzone not impregnated with reagent. They are used to perform tests to see whether the color of a sample (e.g. red juice) stays on the paper of the blank zone. A difference in color can indicate that the intrinsic color of the sample may affect results obtained with other MQuant® test strips.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service