Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.05875

Supelco

Magnesium perchlorate hydrate

[about 83% Mg(ClO₄)₂] for elemental analysis

Synonym(s):

Magnesium perchlorate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(ClO4)2 · x H2O
CAS Number:
Molecular Weight:
223.21 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-108-3
NACRES:
NA.21
form:
solid
solubility:
500 g/L

Quality Level

form

solid

pH

8.2 (25 °C, 50 g/L in H2O)

solubility

500 g/L

density

2.60 g/cm3 at 25 °C (anhydrous substance)

bulk density

600‑800 kg/m3

storage temp.

2-30°C

SMILES string

[Mg+2].[Cl](=O)(=O)(=O)[O-].[Cl](=O)(=O)(=O)[O-].O

InChI

1S/2ClHO4.Mg.H2O/c2*2-1(3,4)5;;/h2*(H,2,3,4,5);;1H2/q;;+2;/p-2

InChI key

KKFFNQXCJMNXHI-UHFFFAOYSA-L

Application


  • Errors in measurements of CO2 with the use of drying agents: Investigates the errors introduced in CO2 measurement techniques using magnesium perchlorate as a drying agent, critical for improving analytical methods in environmental and laboratory settings (Elia et al., 1986).


Analysis Note

Assay (complexometric): ≥ 83 %
Free alkali (as magnesium oxide): ≤ 0.05 %
Free acid (as perchloric acid): ≤ 0.05 %

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service