Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-131-AF488

Sigma-Aldrich

Anti-Sirt1 (Sir2) Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

NAD-dependent protein deacetylase sirtuin-1, Regulatory protein SIR2 homolog 1, SIR2-like protein 1, SIR2alpha, Sir2, mSIR2a, SirtT1 75 kDa fragment, 75SirT1, Sirt1(Sir2), Alexa Fluor 488 Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SIRT1(23411)

General description

NAD-dependent protein deacetylase sirtuin-1 (UniProt Q923E4; also known as Sir2, mSIRT2a, NAD-dependent deacetylase sirtuin-1, Regulatory protein SIR2 homolog 1, Sir2-like 1, SIR2-like protein 1, SIR2alpha) is encoded by the Sirt1 (also known as Sir2l1) gene (Gene ID 93759) in murine species. Sirtuins constitute a family of NAD+-dependent deacetylases found in nearly all organisms. Sirtuins have been implicated in the regulation of aging, transcription, apoptosis, and stress resistance. Full-length SirT1 (FLSirT1) plays an important role in human cartilage homeostasis and chondrocyte survival by mediating the expression of major cartilage anabolic components, collagen 2(I) & aggrecan. The generation of the transcriptionally inactive 75-kDa fragment (75SirT1) via site-specific cleavage of FLSirT1 at amino acid 533 by cathepsin B is an anti-apoptotic mechanism to promote human osteoarthritic (OA) chondrocytes survival following exposure to proinflammatory cytokines. The 5SirT1 fragment is shown to associate with cytochrome c and likely blocks downstream apoptosome assembly. Phosphorylation plays an important role in modulating SirT1 activity and cellular functions. Multiple kinases and their corresponding SirT1 phosphorylation sites have been identified, including pT344 targeted by AMPK, pT530 and pS540 targeted by Cdk1/Cyclin B, pS46 (mouse) or pS47 (human) targeted by JNK1, pT522 targeted by DYRK1A & DYRK3, as well as pS27 & pS47 targeted by CaMKKβ.

Specificity

This antibody recognizes Sirt1.

Immunogen

Epitope: a.a. 1-131
GST-tagged fusion protein corresponding to amino acids 1-131 of mouse Sir2α.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Sirt1(Sir2) in NIH/3T3 cells.
The unconjugated antibody (Cat. No. 07-131) is shown to be suitable also for Western blotting, immunoprecipitation, and ChIP applications.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-Sirt1 (Sir2) Antibody, Alexa Fluor 488 Conjugate is validated for use in Immunocytochemistry for the detection of Sirt1 (Sir2).

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Sirt1(Sir2) in A431 cells.

Target description

~110 kDa observed

Physical form

Affinity purified
Purified rabbit antibody conjugate in PBS with 15 mg/ml BSA and 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service