Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860122W

Avanti

C18:0-d7 GM3 Ceramide (d18:1/18:0-d7)

from synthetic

Synonym(s):

C18:0-d7 GM3 Ganglioside (Neu5Aca2,3Galb1,4Glcb-Ceramide(d18:1/18:0-d7))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H101D7N2O21
CAS Number:
Molecular Weight:
1188.53
UNSPSC Code:
12352211

biological source

synthetic

Assay

≥99% (TLC)

form

methanol solution

packaging

pkg of 1 × 100 μg (860122W-100UG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])CCCCCCCCCCCCCCC(=O)N[C@@H](CO[C@@H]1OC(CO)[C@@H](O[C@@H]2OC(CO)[C@H](O)C(O[C@@]3(CC(O)[C@@H](NC(C)=O)C(O3)[C@H](O)[C@H](O)CO)C(O)=O)C2O)C(O)C1O)C(O)\C=C\CCCCCCCCCCCCC

Application

Lipids research

Packaging

2 mL Amber Glass Sealed Ampule (860122W-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service