Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

840887C

Avanti

22:6 PA

1,2-didocosahexaenoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1,2-di-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphate (sodium salt); PA(22:6(4Z,7Z,10Z,13Z,16Z,19Z)/22:6(4Z,7Z,10Z,13Z,16Z,19Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H68O8PNa
CAS Number:
Molecular Weight:
815.00
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840887C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840887C

concentration

10 mg/mL (840887C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O.[Na+]

InChI

1S/C47H69O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-46(48)53-43-45(44-54-56(50,51)52)55-47(49)42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h5-8,11-14,17-20,23-26,29-32,35-38,45H,3-4,9-10,15-16,21-22,27-28,33-34,39-44H2,1-2H3,(H2,50,51,52);/q;+1/p-1/b7-5-,8-6-,13-11-,14-12-,19-17-,20-18-,25-23-,26-24-,31-29-,32-30-,37-35-,38-36-;/t45-;/m1./s1

InChI key

ZRYYQXRGXSHOSJ-GKSUZLNLSA-M

General description

22:6 PA or 1,2-didocosahexaenoyl-sn-glycero-3-phosphate (sodium salt) is a sodium salt of phosphatidic acid, with substitution of two chains of docosahexaenic acid at the sn-1 and sn-2 positions, in the glycerol backbone.

Application

22:6 PA or 1,2-didocosahexaenoyl-sn-glycero-3-phosphate (sodium salt) has been used as an internal standard in shotgun electrospray ionization tandem mass spectrometry (ESI-MS/MS) for the analysis of phospholipids.

Packaging

5 mL Clear Glass Sealed Ampule (840887C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya Xie et al.
Food chemistry, 285, 194-203 (2019-02-25)
Microwave pretreatment of rapeseeds prior to cold-pressing is a simple and desirable method for producing high quality oils. In this study, a rapid and sensitive lipid profiling platform employing an accurate quantification strategy was established based on direct infusion electrospray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service